Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297649.1 | NP_001284578.1 | 3 - 4 | 393 | 105 |
NM_004356.3 | NP_004347.1 | 3 - 4 | 511 | 105 | |
XM_011520492.2 | XP_011518794.1 | 3 - 4 | 745 | 105 | |
XM_017018598.1 | XP_016874087.1 | 3 - 4 | 254 | 105 | |
GenBank mRNA | AB209380.1 | - | 1 - 2 | 2809 | 105 |
BC002978.1 | - | 3 - 4 | 318 | 105 | |
BC093047.1 | - | 3 - 4 | 509 | 105 | |
BG397506.1 | - | 4 - 5 | 381 | 105 | |
BP323362.1 | - | 3 - 4 | 407 | 105 | |
BQ709484.1 | - | - | 508 | 105 | |
BT019507.1 | - | 3 - 4 | 278 | 105 | |
BT019508.1 | - | 3 - 4 | 278 | 105 | |
M33680.1 | - | 3 - 4 | 516 | 105 |