Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297649.1 | NP_001284578.1 | 4 - 5 | 470 | 67 |
NM_004356.3 | NP_004347.1 | 4 - 5 | 588 | 67 | |
XM_011520492.2 | XP_011518794.1 | 4 - 5 | 822 | 67 | |
XM_017018598.1 | XP_016874087.1 | 4 - 5 | 331 | 67 | |
GenBank mRNA | BC002978.1 | - | 4 - 5 | 395 | 67 |
BC093047.1 | - | 4 - 5 | 586 | 67 | |
BG397506.1 | - | 5 - 6 | 458 | 67 | |
BP323362.1 | - | 4 - 5 | 484 | 67 | |
BQ709484.1 | - | - | 585 | 67 | |
BT019507.1 | - | 4 - 5 | 355 | 67 | |
BT019508.1 | - | 4 - 5 | 355 | 67 | |
M33680.1 | - | 4 - 5 | 593 | 67 |