Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014981.1 | NP_055796.1 | 11 - 12 | 1099 | 99 |
XM_011512559.2 | XP_011510861.1 | 12 - 13 | 1220 | 99 | |
XM_017005922.1 | XP_016861411.1 | 2 - 3 | 364 | 99 | |
GenBank mRNA | AB023217.3 | - | 11 - 12 | 1099 | 99 |