Assay Design



Gene Transcripts

Gene Symbol
BAZ2B
Entrez Gene ID
29994
Gene Name
bromodomain adjacent to zinc finger domain 2B
Gene Aliases
WALp4
Chromosome Location
Chr.2: 159315312 - 159712439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289975.1 NP_001276904.1 27 - 28 5162 82
NM_013450.3 NP_038478.2 28 - 29 5270 82
XM_005246488.2 XP_005246545.2 29 - 30 5146 82
XM_005246489.4 XP_005246546.2 28 - 29 6249 82
XM_005246492.4 XP_005246549.2 28 - 29 6244 82
XM_006712461.3 XP_006712524.2 32 - 33 5720 82
XM_011511038.2 XP_011509340.1 29 - 30 5244 82
XM_011511039.1 XP_011509341.1 26 - 27 4867 82
XM_011511042.1 XP_011509344.1 27 - 28 4900 82
XM_011511043.1 XP_011509345.1 25 - 26 4765 82
XM_011511044.1 XP_011509346.1 26 - 27 4792 82
XM_011511045.1 XP_011509347.1 26 - 27 4705 82
XM_011511047.1 XP_011509349.1 28 - 29 5052 82
XM_011511048.2 XP_011509350.1 28 - 29 5117 82
XM_011511050.1 XP_011509352.1 26 - 27 4600 82
XM_011511051.1 XP_011509353.1 26 - 27 4786 82
XM_011511052.1 XP_011509354.1 25 - 26 4504 82
XM_011511053.2 XP_011509355.1 27 - 28 4900 82
XM_011511054.2 XP_011509356.1 27 - 28 4900 82
XM_011511056.1 XP_011509358.1 17 - 18 2520 82
XM_017003918.1 XP_016859407.1 32 - 33 5710 82
XM_017003919.1 XP_016859408.1 32 - 33 5704 82
XM_017003920.1 XP_016859409.1 34 - 35 5921 82
XM_017003921.1 XP_016859410.1 29 - 30 5244 82
XM_017003922.1 XP_016859411.1 29 - 30 5140 82
XM_017003923.1 XP_016859412.1 27 - 28 4894 82
XM_017003924.1 XP_016859413.1 26 - 27 4792 82
XM_017003925.1 XP_016859414.1 28 - 29 5148 82
XM_017003926.1 XP_016859415.1 28 - 29 5006 82
XM_017003927.1 XP_016859416.1 28 - 29 5046 82
XM_017003928.1 XP_016859417.1 28 - 29 4956 82
XM_017003929.1 XP_016859418.1 28 - 29 5038 82
XM_017003930.1 XP_016859419.1 28 - 29 4846 82
XM_017003931.1 XP_016859420.1 28 - 29 4944 82
XM_017003932.1 XP_016859421.1 25 - 26 4597 82
XM_017003933.1 XP_016859422.1 27 - 28 4823 82
XM_017003934.1 XP_016859423.1 27 - 28 4718 82
XM_017003935.1 XP_016859424.1 26 - 27 4656 82
XM_017003936.1 XP_016859425.1 26 - 27 4610 82
XM_017003937.1 XP_016859426.1 21 - 22 3317 82
GenBank mRNA AB032255.1 - 23 - 24 4643 82
AB040909.2 - 27 - 28 5130 82
AK074895.1 - 6 - 7 1388 82

View Full Product Details