Assay Design



Gene Transcripts

Gene Symbol
BAZ2B
Entrez Gene ID
29994
Gene Name
bromodomain adjacent to zinc finger domain 2B
Gene Aliases
WALp4
Chromosome Location
Chr.2: 159315312 - 159712439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289975.1 NP_001276904.1 33 - 34 6245 107
NM_013450.3 NP_038478.2 34 - 35 6353 107
XM_005246488.2 XP_005246545.2 35 - 36 6334 107
XM_005246489.4 XP_005246546.2 34 - 35 7437 107
XM_005246492.4 XP_005246549.2 34 - 35 7432 107
XM_006712461.3 XP_006712524.2 38 - 39 6908 107
XM_011511038.2 XP_011509340.1 35 - 36 6432 107
XM_011511039.1 XP_011509341.1 32 - 33 6055 107
XM_011511042.1 XP_011509344.1 33 - 34 5983 107
XM_011511043.1 XP_011509345.1 31 - 32 5953 107
XM_011511044.1 XP_011509346.1 32 - 33 5980 107
XM_011511045.1 XP_011509347.1 32 - 33 5893 107
XM_011511047.1 XP_011509349.1 34 - 35 6240 107
XM_011511048.2 XP_011509350.1 34 - 35 6305 107
XM_011511050.1 XP_011509352.1 32 - 33 5788 107
XM_011511051.1 XP_011509353.1 32 - 33 5869 107
XM_011511052.1 XP_011509354.1 31 - 32 5692 107
XM_011511056.1 XP_011509358.1 23 - 24 3708 107
XM_017003918.1 XP_016859407.1 38 - 39 6898 107
XM_017003919.1 XP_016859408.1 38 - 39 6892 107
XM_017003920.1 XP_016859409.1 40 - 41 7109 107
XM_017003921.1 XP_016859410.1 35 - 36 6432 107
XM_017003922.1 XP_016859411.1 35 - 36 6328 107
XM_017003923.1 XP_016859412.1 33 - 34 6082 107
XM_017003924.1 XP_016859413.1 32 - 33 5980 107
XM_017003925.1 XP_016859414.1 34 - 35 6336 107
XM_017003926.1 XP_016859415.1 34 - 35 6194 107
XM_017003927.1 XP_016859416.1 34 - 35 6234 107
XM_017003928.1 XP_016859417.1 34 - 35 6144 107
XM_017003929.1 XP_016859418.1 34 - 35 6121 107
XM_017003930.1 XP_016859419.1 34 - 35 6034 107
XM_017003931.1 XP_016859420.1 34 - 35 6132 107
XM_017003932.1 XP_016859421.1 31 - 32 5785 107
XM_017003933.1 XP_016859422.1 33 - 34 6011 107
XM_017003934.1 XP_016859423.1 33 - 34 5906 107
XM_017003935.1 XP_016859424.1 32 - 33 5844 107
XM_017003936.1 XP_016859425.1 32 - 33 5798 107
XM_017003937.1 XP_016859426.1 27 - 28 4505 107
GenBank mRNA AB040909.2 - 33 - 34 6213 107
AK074895.1 - 12 - 13 2471 107
AL080173.1 - 5 - 6 793 107

View Full Product Details