Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190266.1 | NP_001177195.1 | 6 - 7 | 955 | 113 |
NM_001190267.1 | NP_001177196.1 | 6 - 7 | 953 | 113 | |
NM_017974.3 | NP_060444.3 | 6 - 7 | 957 | 113 | |
NM_030803.6 | NP_110430.5 | 6 - 7 | 957 | 113 | |
NM_198890.2 | NP_942593.2 | 3 - 4 | 525 | 113 | |
XM_005246082.2 | XP_005246139.1 | 6 - 7 | 894 | 113 | |
XM_005246084.2 | XP_005246141.1 | 3 - 4 | 433 | 113 | |
XM_005246086.2 | XP_005246143.1 | 3 - 4 | 433 | 113 | |
XM_006712608.2 | XP_006712671.1 | 5 - 6 | 705 | 113 | |
GenBank mRNA | AK001690.1 | - | 6 - 7 | 803 | 113 |
AK024453.1 | - | 6 - 7 | 827 | 113 | |
AK027854.1 | - | 5 - 6 | 539 | 113 | |
AK123876.1 | - | 3 - 4 | 424 | 113 | |
AY358182.1 | - | 6 - 7 | 846 | 113 | |
AY398617.1 | - | 6 - 7 | 846 | 113 | |
BC071846.1 | - | 3 - 4 | 363 | 113 | |
BC117337.1 | - | 6 - 7 | 619 | 113 | |
BC143900.1 | - | 6 - 7 | 619 | 113 | |
EF079889.1 | - | 6 - 7 | 700 | 113 | |
EF079890.1 | - | 3 - 4 | 268 | 113 | |
EF560734.1 | - | 6 - 7 | 827 | 113 | |
JQ924062.1 | - | 6 - 7 | 700 | 113 |