Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012417.3 | NP_036549.2 | 6 - 7 | 1333 | 70 |
NM_181671.2 | NP_858057.1 | 6 - 7 | 1333 | 70 | |
XM_005257216.2 | XP_005257273.1 | 6 - 7 | 833 | 70 | |
XM_017024442.1 | XP_016879931.1 | 6 - 7 | 444 | 70 | |
XM_017024443.1 | XP_016879932.1 | 6 - 7 | 444 | 70 | |
XM_017024444.1 | XP_016879933.1 | 6 - 7 | 835 | 70 | |
XM_017024445.1 | XP_016879934.1 | 6 - 7 | 457 | 70 | |
GenBank mRNA | AK290838.1 | - | 6 - 7 | 1155 | 70 |
AK307138.1 | - | 3 - 4 | 597 | 70 | |
BC007905.2 | - | 6 - 7 | 806 | 70 | |
BC067095.1 | - | 6 - 7 | 1054 | 70 | |
BE906407.1 | - | 4 - 5 | 239 | 70 |