Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021229.3 | NP_067052.2 | 3 - 4 | 1307 | 126 |
GenBank mRNA | AF119916.3 | - | 4 - 5 | 844 | 126 |
AF278532.1 | - | 3 - 4 | 1309 | 126 | |
AK024691.1 | - | 2 - 3 | 383 | 126 | |
AK226070.1 | - | 3 - 4 | 1164 | 126 | |
AK290591.1 | - | 3 - 4 | 844 | 126 | |
AK315796.1 | - | 3 - 4 | 1166 | 126 | |
AL137540.1 | - | 1 - 2 | 175 | 126 | |
AL833767.1 | - | 2 - 3 | 711 | 126 | |
AY330211.1 | - | 3 - 4 | 858 | 126 | |
BC013591.1 | - | 2 - 3 | 382 | 126 | |
BC136798.1 | - | 3 - 4 | 1282 | 126 | |
BC136799.1 | - | 3 - 4 | 1282 | 126 |