Assay Design



Gene Transcripts

Gene Symbol
NTN4
Entrez Gene ID
59277
Gene Name
netrin 4
Gene Aliases
PRO3091
Chromosome Location
Chr.12: 95657807 - 95790758 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021229.3 NP_067052.2 4 - 5 1443 73
GenBank mRNA AF119916.3 - 5 - 6 980 73
AF297711.1 - 4 - 5 1295 73
AK024691.1 - 3 - 4 519 73
AK226070.1 - 4 - 5 1300 73
AK290591.1 - 4 - 5 980 73
AK315796.1 - 4 - 5 1302 73
AL137540.1 - 2 - 3 311 73
AL833767.1 - 3 - 4 847 73
BC013591.1 - 3 - 4 518 73
BC136798.1 - 4 - 5 1418 73
BC136799.1 - 4 - 5 1418 73

View Full Product Details