Assay Design



Gene Transcripts

Gene Symbol
SLC35F2
Entrez Gene ID
54733
Gene Name
solute carrier family 35 member F2
Gene Aliases
HSNOV1
Chromosome Location
Chr.11: 107790991 - 107859188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017515.4 NP_059985.2 7 - 8 1352 80
GenBank mRNA AK023080.1 - 7 - 8 949 80
AK300582.1 - 6 - 7 787 80
BC039195.2 - 7 - 8 1092 80
BC048302.1 - 7 - 8 958 80

View Full Product Details