Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017670.2 | NP_060140.2 | 4 - 5 | 940 | 64 |
GenBank mRNA | AF161381.1 | - | 4 - 5 | 348 | 64 |
AK000120.1 | - | 4 - 5 | 380 | 64 | |
AK225133.1 | - | 4 - 5 | 347 | 64 | |
AK298287.1 | - | 5 - 6 | 659 | 64 | |
AK307413.1 | - | 4 - 5 | 475 | 64 | |
AY177200.1 | - | 4 - 5 | 336 | 64 | |
BC007519.2 | - | 4 - 5 | 344 | 64 | |
BC010368.1 | - | 4 - 5 | 366 | 64 | |
BC107701.1 | - | 4 - 5 | 366 | 64 |