Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172746.1 | NP_001166217.1 | 1 - 2 | 423 | 93 |
NM_032985.4 | NP_116780.1 | 1 - 2 | 445 | 93 | |
GenBank mRNA | AK296270.1 | - | 1 - 2 | 99 | 93 |
AK299513.1 | - | 1 - 2 | 49 | 93 | |
BC005032.1 | - | 1 - 2 | 87 | 93 | |
BP244573.1 | - | 1 - 2 | 452 | 93 |