Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019593.3 | NP_062539.1 | 15 - 16 | 1536 | 126 |
XM_005260758.4 | XP_005260815.1 | 16 - 17 | 1687 | 126 | |
XM_005260761.1 | XP_005260818.1 | 14 - 15 | 1460 | 126 | |
XM_006723596.3 | XP_006723659.1 | 16 - 17 | 2460 | 126 | |
GenBank mRNA | AB037855.1 | - | 15 - 16 | 1527 | 126 |
AK001947.1 | - | 2 - 3 | 89 | 126 | |
AL833069.1 | - | 10 - 11 | 2732 | 126 | |
BC027588.2 | - | 15 - 16 | 1535 | 126 |