Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019593.3 | NP_062539.1 | 3 - 4 | 357 | 78 |
XM_005260758.4 | XP_005260815.1 | 4 - 5 | 508 | 78 | |
XM_005260761.1 | XP_005260818.1 | 3 - 4 | 357 | 78 | |
XM_006723596.3 | XP_006723659.1 | 4 - 5 | 1281 | 78 | |
GenBank mRNA | AB037855.1 | - | 3 - 4 | 348 | 78 |
AK308111.1 | - | 3 - 4 | 355 | 78 | |
BC027588.2 | - | 3 - 4 | 356 | 78 | |
BI561344.1 | - | 3 - 4 | 290 | 78 |