Assay Design



Gene Transcripts

Gene Symbol
CAPN9
Entrez Gene ID
10753
Gene Name
calpain 9
Gene Aliases
GC36, nCL-4
Chromosome Location
Chr.1: 230747289 - 230803356 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319676.1 NP_001306605.1 14 - 15 1707 85
NM_006615.2 NP_006606.1 16 - 17 1896 85
NM_016452.2 NP_057536.1 15 - 16 1818 85
XM_011544017.1 XP_011542319.1 16 - 17 1880 85
XM_011544018.1 XP_011542320.1 15 - 16 1802 85
XM_011544019.2 XP_011542321.1 14 - 15 1802 85
XM_011544020.1 XP_011542322.1 16 - 17 1626 85
XM_017000098.1 XP_016855587.1 15 - 16 1796 85
XM_017000099.1 XP_016855588.1 16 - 17 1626 85
GenBank mRNA AB038463.1 - 15 - 16 1787 85
AF022799.1 - 16 - 17 1896 85
AK298024.1 - 14 - 15 1646 85
AK313629.1 - 16 - 17 1880 85
BC027993.1 - 14 - 15 1658 85

View Full Product Details