Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319676.1 | NP_001306605.1 | 7 - 8 | 1036 | 117 |
NM_006615.2 | NP_006606.1 | 9 - 10 | 1225 | 117 | |
NM_016452.2 | NP_057536.1 | 8 - 9 | 1147 | 117 | |
XM_011544017.1 | XP_011542319.1 | 9 - 10 | 1209 | 117 | |
XM_011544018.1 | XP_011542320.1 | 8 - 9 | 1131 | 117 | |
XM_011544019.2 | XP_011542321.1 | 7 - 8 | 1131 | 117 | |
XM_011544020.1 | XP_011542322.1 | 9 - 10 | 955 | 117 | |
XM_017000098.1 | XP_016855587.1 | 8 - 9 | 1125 | 117 | |
XM_017000099.1 | XP_016855588.1 | 9 - 10 | 955 | 117 | |
GenBank mRNA | AB038463.1 | - | 8 - 9 | 1116 | 117 |
AF022799.1 | - | 9 - 10 | 1225 | 117 | |
AK298024.1 | - | 7 - 8 | 975 | 117 | |
AK309039.1 | - | 9 - 10 | 1021 | 117 | |
AK313629.1 | - | 9 - 10 | 1209 | 117 | |
BC027993.1 | - | 7 - 8 | 987 | 117 |