Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017425.2 | NP_001017425.2 | 1 - 2 | 215 | 68 |
GenBank mRNA | AF004711.1 | - | 1 - 2 | 199 | 68 |
AK291483.1 | - | 1 - 2 | 195 | 68 | |
AK294780.1 | - | 1 - 2 | 197 | 68 | |
DR001358.1 | - | 1 - 2 | 224 | 68 | |
EF165334.1 | - | 1 - 2 | 148 | 68 | |
EF165335.1 | - | 1 - 2 | 113 | 68 |