Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030630.2 | NP_085133.1 | 14 - 15 | 2011 | 59 |
XM_005257226.1 | XP_005257283.1 | 14 - 15 | 2006 | 59 | |
GenBank mRNA | AK074401.1 | - | 14 - 15 | 1939 | 59 |
AK075111.1 | - | 4 - 5 | 536 | 59 | |
AK125514.1 | - | 1 - 2 | 832 | 59 | |
AK304081.1 | - | 14 - 15 | 1895 | 59 | |
BC032219.1 | - | 10 - 11 | 1309 | 59 | |
BC035372.1 | - | 14 - 15 | 1927 | 59 | |
BX496609.1 | - | 2 - 3 | 383 | 59 |