Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017780.3 | NP_060250.2 | 7 - 8 | 2987 | 141 |
XM_011517553.2 | XP_011515855.1 | 7 - 8 | 3008 | 141 | |
XM_011517554.2 | XP_011515856.1 | 7 - 8 | 2986 | 141 | |
XM_011517555.2 | XP_011515857.1 | 7 - 8 | 3008 | 141 | |
XM_011517560.2 | XP_011515862.1 | 7 - 8 | 3008 | 141 | |
XM_017013612.1 | XP_016869101.1 | 7 - 8 | 3062 | 141 | |
XM_017013613.1 | XP_016869102.1 | 7 - 8 | 3008 | 141 | |
GenBank mRNA | AB037837.1 | - | 3 - 4 | 209 | 141 |