Assay Design
Gene Transcripts
- Gene Symbol
- FGFR3
- Entrez Gene ID
-
2261
- Gene Name
- fibroblast growth factor receptor 3
- Gene Aliases
-
ACH, CD333, CEK2, HSFGFR3EX, JTK4
- Chromosome Location
-
Chr.4: 1793299 - 1808872 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_022965.3 |
NP_075254.1 |
7 - 8 |
1184 |
138 |
GenBank mRNA |
AF245114.1 |
- |
6 - 7 |
967 |
138 |
View Full Product Details