Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020749.4 | NP_065800.1 | 1 - 2 | 485 | 78 |
GenBank mRNA | AF121259.1 | - | 1 - 2 | 125 | 78 |
AF293357.1 | - | 1 - 2 | 398 | 78 | |
AK314692.1 | - | 1 - 2 | 419 | 78 | |
BC117564.1 | - | 1 - 2 | 163 | 78 | |
BC142971.1 | - | 1 - 2 | 418 | 78 |