Assay Design
Gene Transcripts
- Gene Symbol
- OR13C5
- Entrez Gene ID
-
138799
- Gene Name
- olfactory receptor family 13 subfamily C member 5
- Gene Aliases
-
OR9-11
- Chromosome Location
-
Chr.9: 104598457 - 104599413 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC136843.1 |
- |
1 - 1 |
1055 |
161 |
View Full Product Details