Assay Design
Gene Transcripts
- Gene Symbol
- TATDN2
- Entrez Gene ID
-
9797
- Gene Name
- TatD DNase domain containing 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 10248493 - 10281222 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_014760.3 |
NP_055575.3 |
7 - 8 |
2908 |
59 |
GenBank mRNA |
BC090935.1 |
- |
7 - 8 |
2489 |
59 |
View Full Product Details