Assay Design
Gene Transcripts
- Gene Symbol
- NETO1
- Entrez Gene ID
-
81832
- Gene Name
- neuropilin and tolloid like 1
- Gene Aliases
-
BCTL1, BTCL1
- Chromosome Location
-
Chr.18: 72742314 - 72867667 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138999.1 |
NP_620552.1 |
1 - 2 |
128 |
88 |
GenBank mRNA |
AF448839.1 |
- |
1 - 2 |
128 |
88 |
View Full Product Details