Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297595.1 | NP_001284524.1 | 16 - 17 | 2890 | 102 |
NM_001297597.1 | NP_001284526.1 | 7 - 8 | 1996 | 102 | |
NM_015260.3 | NP_056075.1 | 17 - 18 | 2986 | 102 | |
XM_006722704.1 | XP_006722767.1 | 7 - 8 | 1601 | 102 | |
GenBank mRNA | AB014600.2 | - | 16 - 17 | 2877 | 102 |
AK001633.1 | - | 10 - 11 | 1991 | 102 | |
AK295618.1 | - | 7 - 8 | 1935 | 102 | |
AY706204.1 | - | 17 - 18 | 2966 | 102 | |
BC005113.1 | - | 4 - 5 | 728 | 102 |