Assay Design
Gene Transcripts
- Gene Symbol
- SYNGAP1
- Entrez Gene ID
-
8831
- Gene Name
- synaptic Ras GTPase activating protein 1
- Gene Aliases
-
MRD5, RASA1, RASA5, SYNGAP
- Chromosome Location
-
Chr.6: 33420070 - 33453689 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_006772.2 |
NP_006763.2 |
18 - 19 |
4077 |
78 |
GenBank mRNA |
BX282051.1 |
- |
3 - 4 |
468 |
78 |
View Full Product Details