Assay Design



Gene Transcripts

Gene Symbol
BRD4
Entrez Gene ID
23476
Gene Name
bromodomain containing 4
Gene Aliases
CAP, HUNK1, HUNKI, MCAP
Chromosome Location
Chr.19: 15236836 - 15332543 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058243.2 NP_490597.1 14 - 15 3393 100
XM_011527854.1 XP_011526156.1 14 - 15 3458 100
GenBank mRNA AF386649.1 - 14 - 15 3393 100

View Full Product Details