Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079843.2 | NP_001073312.1 | 6 - 7 | 1684 | 84 |
NM_017766.4 | NP_060236.3 | 6 - 7 | 1684 | 84 | |
XM_005263479.3 | XP_005263536.1 | 4 - 5 | 1548 | 84 | |
XM_011541635.2 | XP_011539937.1 | 4 - 5 | 1547 | 84 | |
XM_017001539.1 | XP_016857028.1 | 4 - 5 | 1548 | 84 | |
XM_017001540.1 | XP_016857029.1 | 6 - 7 | 1658 | 84 | |
XM_017001541.1 | XP_016857030.1 | 6 - 7 | 1720 | 84 | |
GenBank mRNA | AK000328.1 | - | 2 - 3 | 998 | 84 |
AK022285.1 | - | 1 - 2 | 1695 | 84 | |
AK092289.1 | - | 6 - 7 | 1658 | 84 | |
AK225324.1 | - | 2 - 3 | 998 | 84 | |
BC051883.2 | - | 3 - 4 | 1354 | 84 | |
DQ217660.1 | - | 6 - 7 | 1656 | 84 |