Assay Design



Gene Transcripts

Gene Symbol
RUNX1T1
Entrez Gene ID
862
Gene Name
RUNX1 translocation partner 1
Gene Aliases
AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
Chromosome Location
Chr.8: 91954967 - 92103365 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198633.1 NP_001185562.1 1 - 2 371 60
GenBank mRNA DA779230.1 - 1 - 2 332 60

View Full Product Details