Assay Design



Gene Transcripts

Gene Symbol
DIP2A
Entrez Gene ID
23181
Gene Name
disco interacting protein 2 homolog A
Gene Aliases
C21orf106, DIP2
Chromosome Location
Chr.21: 46458942 - 46570900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146116.1 NP_001139588.1 6 - 7 970 55
NM_015151.3 NP_055966.2 6 - 7 970 55
NM_206889.2 NP_996772.1 6 - 7 970 55
NM_206890.2 NP_996773.1 6 - 7 970 55
NM_206891.2 NP_996774.1 6 - 7 970 55
XM_011529492.2 XP_011527794.1 7 - 8 1085 55
XM_011529501.1 XP_011527803.1 7 - 8 1085 55
XM_011529502.1 XP_011527804.1 7 - 8 1085 55
XM_017028291.1 XP_016883780.1 7 - 8 1085 55
XM_017028292.1 XP_016883781.1 7 - 8 1085 55
XM_017028293.1 XP_016883782.1 7 - 8 1082 55
XM_017028294.1 XP_016883783.1 6 - 7 964 55
XM_017028295.1 XP_016883784.1 6 - 7 961 55
XM_017028296.1 XP_016883785.1 6 - 7 961 55
XM_017028299.1 XP_016883788.1 6 - 7 830 55
XM_017028300.1 XP_016883789.1 7 - 8 1085 55
XM_017028301.1 XP_016883790.1 7 - 8 1085 55
XM_017028302.1 XP_016883791.1 5 - 6 725 55
XM_017028303.1 XP_016883792.1 7 - 8 1085 55
XM_017028304.1 XP_016883793.1 7 - 8 1085 55
XM_017028305.1 XP_016883794.1 7 - 8 1085 55
GenBank mRNA AB273729.1 - 6 - 7 808 55
AF432263.1 - 1 - 2 180 55
AF490768.1 - 6 - 7 875 55
AK056738.1 - 6 - 7 862 55
AK098236.1 - 1 - 2 113 55
AK303351.1 - 4 - 5 785 55
BC038443.1 - 6 - 7 969 55

View Full Product Details