Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004984.2 | NP_004975.2 | 24 - 25 | 2964 | 78 |
GenBank mRNA | AB210045.1 | - | 24 - 25 | 2789 | 78 |
AB290174.1 | - | 24 - 25 | 3030 | 78 | |
AK294881.1 | - | 21 - 22 | 2717 | 78 | |
AK315202.1 | - | 24 - 25 | 2984 | 78 | |
BC146670.1 | - | 24 - 25 | 2967 | 78 | |
BC150208.1 | - | 24 - 25 | 2967 | 78 | |
BM930000.1 | - | 1 - 2 | 160 | 78 | |
U06698.1 | - | 24 - 25 | 2904 | 78 |