Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018043.5 | NP_060513.5 | 2 - 3 | 748 | 116 |
NR_030691.1 | 2 - 3 | 596 | 116 | ||
XM_006718602.2 | XP_006718665.1 | 2 - 3 | 464 | 116 | |
XM_006718604.2 | XP_006718667.1 | 2 - 3 | 579 | 116 | |
XM_006718605.2 | XP_006718668.1 | 2 - 3 | 591 | 116 | |
XM_011545121.2 | XP_011543423.1 | 2 - 3 | 456 | 116 | |
XM_011545123.2 | XP_011543425.1 | 2 - 3 | 456 | 116 | |
XM_011545124.2 | XP_011543426.1 | 2 - 3 | 456 | 116 | |
XM_011545125.2 | XP_011543427.1 | 2 - 3 | 456 | 116 | |
XM_011545126.2 | XP_011543428.1 | 2 - 3 | 456 | 116 | |
XM_011545127.2 | XP_011543429.1 | 2 - 3 | 456 | 116 | |
XM_011545128.2 | XP_011543430.1 | 2 - 3 | 456 | 116 | |
XM_011545129.2 | XP_011543431.1 | 2 - 3 | 371 | 116 | |
XM_011545131.2 | XP_011543433.1 | 2 - 3 | 464 | 116 | |
XM_017017956.1 | XP_016873445.1 | 2 - 3 | 456 | 116 | |
XM_017017957.1 | XP_016873446.1 | 2 - 3 | 456 | 116 | |
GenBank mRNA | AB845669.1 | - | 2 - 3 | 443 | 116 |
AK097619.1 | - | 2 - 3 | 464 | 116 | |
AK293088.1 | - | 2 - 3 | 456 | 116 | |
AL833582.1 | - | 2 - 3 | 492 | 116 | |
AY728143.1 | - | 2 - 3 | 748 | 116 | |
BC033036.2 | - | 2 - 3 | 552 | 116 | |
CF593428.1 | - | 2 - 3 | 538 | 116 |