Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018043.5 | NP_060513.5 | 6 - 7 | 1105 | 66 |
NR_030691.1 | 6 - 7 | 953 | 66 | ||
XM_006718605.2 | XP_006718668.1 | 6 - 7 | 948 | 66 | |
XM_011545127.2 | XP_011543429.1 | 6 - 7 | 813 | 66 | |
XM_011545128.2 | XP_011543430.1 | 6 - 7 | 813 | 66 | |
XM_011545131.2 | XP_011543433.1 | 6 - 7 | 821 | 66 | |
XM_017017956.1 | XP_016873445.1 | 6 - 7 | 813 | 66 | |
XM_017017957.1 | XP_016873446.1 | 6 - 7 | 813 | 66 | |
GenBank mRNA | AB845669.1 | - | 6 - 7 | 800 | 66 |
AK097619.1 | - | 6 - 7 | 821 | 66 | |
AK293088.1 | - | 6 - 7 | 813 | 66 | |
AL833582.1 | - | 6 - 7 | 849 | 66 | |
AY728143.1 | - | 6 - 7 | 1105 | 66 | |
BC027590.2 | - | 1 - 2 | 57 | 66 | |
BC033036.2 | - | 6 - 7 | 909 | 66 |