Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002660.2 | NP_002651.2 | 30 - 31 | 3771 | 73 |
XM_005260438.2 | XP_005260495.1 | 29 - 30 | 3890 | 73 | |
XM_011528865.1 | XP_011527167.1 | 29 - 30 | 3755 | 73 | |
GenBank mRNA | AK297370.1 | - | 18 - 19 | 2638 | 73 |
AL110247.1 | - | 12 - 13 | 1505 | 73 | |
BC144136.1 | - | 30 - 31 | 3738 | 73 |