Assay Design



Gene Transcripts

Gene Symbol
ITCH
Entrez Gene ID
83737
Gene Name
itchy E3 ubiquitin protein ligase
Gene Aliases
ADMFD, AIF4, AIP4, NAPP1
Chromosome Location
Chr.20: 34363235 - 34511773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324197.1 NP_001311126.1 1 - 2 117 152
NM_001324198.1 NP_001311127.1 1 - 2 117 152

View Full Product Details