Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018327.2 | NP_060797.2 | 3 - 4 | 747 | 116 |
XM_011529279.1 | XP_011527581.1 | 3 - 4 | 515 | 116 | |
XM_017027922.1 | XP_016883411.1 | 3 - 4 | 515 | 116 | |
GenBank mRNA | AK300184.1 | - | 3 - 4 | 553 | 116 |
BC150644.1 | - | 3 - 4 | 706 | 116 | |
CF994231.1 | - | 3 - 4 | 521 | 116 | |
CX870585.1 | - | 2 - 3 | 240 | 116 |