Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005870.4 | NP_005861.2 | 1 - 2 | 165 | 74 |
GenBank mRNA | AF153608.1 | - | 1 - 2 | 103 | 74 |
AK311748.1 | - | 1 - 2 | 86 | 74 | |
AY550970.1 | - | 1 - 2 | 85 | 74 | |
BC030836.1 | - | 1 - 2 | 133 | 74 | |
U78303.1 | - | 1 - 2 | 100 | 74 | |
U96915.1 | - | 1 - 2 | 85 | 74 |