Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190415.1 | NP_001177344.1 | 10 - 11 | 1310 | 57 |
NM_005641.3 | NP_005632.1 | 10 - 11 | 1639 | 57 | |
NM_139315.2 | NP_647476.1 | 10 - 11 | 1341 | 57 | |
NR_033792.1 | 10 - 11 | 1190 | 57 | ||
XM_006716100.2 | XP_006716163.1 | 11 - 12 | 1432 | 57 | |
XM_006716101.2 | XP_006716164.1 | 10 - 11 | 1269 | 57 | |
XM_006716102.2 | XP_006716165.1 | 10 - 11 | 1136 | 57 | |
XM_011516542.2 | XP_011514844.1 | 11 - 12 | 1411 | 57 | |
XM_011516543.2 | XP_011514845.1 | 10 - 11 | 1164 | 57 | |
XM_017012568.1 | XP_016868057.1 | 10 - 11 | 1609 | 57 | |
GenBank mRNA | AK097781.1 | - | 10 - 11 | 1192 | 57 |
AK300005.1 | - | 10 - 11 | 1293 | 57 | |
AK314428.1 | - | 10 - 11 | 1616 | 57 | |
AK315077.1 | - | 10 - 11 | 1291 | 57 | |
BC018115.1 | - | 10 - 11 | 1284 | 57 | |
CR627390.1 | - | 10 - 11 | 1168 | 57 | |
L25444.1 | - | 10 - 11 | 1611 | 57 | |
U31659.1 | - | 9 - 10 | 1395 | 57 |