Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173165.2 | NP_775188.1 | 9 - 10 | 3426 | 75 |
GenBank mRNA | AK290370.1 | - | 9 - 10 | 3166 | 75 |
BC001050.2 | - | 9 - 10 | 3144 | 75 | |
EU887606.1 | - | 9 - 10 | 3110 | 75 | |
EU887609.1 | - | 9 - 10 | 3437 | 75 | |
EU887612.1 | - | 9 - 10 | 3155 | 75 | |
EU887615.1 | - | 9 - 10 | 3063 | 75 | |
EU887618.1 | - | 9 - 10 | 3068 | 75 | |
EU887621.1 | - | 9 - 10 | 3105 | 75 | |
EU887624.1 | - | 9 - 10 | 3509 | 75 | |
U14510.1 | - | 9 - 10 | 3134 | 75 |