Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083907.2 | NP_001077376.2 | 13 - 14 | 2258 | 73 |
NM_001127507.2 | NP_001120979.2 | 12 - 13 | 2090 | 73 | |
NM_017935.4 | NP_060405.4 | 13 - 14 | 2489 | 73 | |
XM_017008337.1 | XP_016863826.1 | 13 - 14 | 2236 | 73 | |
GenBank mRNA | AB063170.1 | - | 12 - 13 | 2125 | 73 |
AK000713.1 | - | 13 - 14 | 2207 | 73 | |
AK091523.1 | - | 17 - 18 | 2590 | 73 | |
AK093024.1 | - | 1 - 2 | 1755 | 73 | |
AK292133.1 | - | 13 - 14 | 2333 | 73 | |
AL834291.1 | - | 13 - 14 | 2308 | 73 | |
BC032241.1 | - | 13 - 14 | 2357 | 73 | |
EU051376.1 | - | 12 - 13 | 1828 | 73 |