Assay Design



Gene Transcripts

Gene Symbol
BANK1
Entrez Gene ID
55024
Gene Name
B-cell scaffold protein with ankyrin repeats 1
Gene Aliases
BANK
Chromosome Location
Chr.4: 101790482 - 102074812 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083907.2 NP_001077376.2 1 - 2 104 148
GenBank mRNA AK000713.1 - 1 - 2 51 148
DA946668.1 - 1 - 2 104 149

View Full Product Details