Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152592.3 | NP_689805.3 | 6 - 7 | 1285 | 90 |
XM_005267377.3 | XP_005267434.1 | 7 - 8 | 1382 | 90 | |
XM_006720063.3 | XP_006720126.1 | 7 - 8 | 1482 | 90 | |
XM_011536515.2 | XP_011534817.1 | 7 - 8 | 1417 | 90 | |
XM_017021056.1 | XP_016876545.1 | 6 - 7 | 1284 | 90 | |
XM_017021057.1 | XP_016876546.1 | 7 - 8 | 1382 | 90 | |
GenBank mRNA | AK131436.1 | - | 7 - 8 | 1150 | 90 |
BC146604.1 | - | 7 - 8 | 1285 | 90 | |
BX248076.1 | - | 6 - 7 | 1285 | 90 | |
BX342493.1 | - | 3 - 4 | 587 | 90 |