Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130037.1 | NP_001123509.1 | 2 - 3 | 420 | 84 |
NM_018712.3 | NP_061182.3 | 2 - 3 | 420 | 84 | |
GenBank mRNA | AK054794.1 | - | 2 - 3 | 240 | 84 |
AK303689.1 | - | 2 - 3 | 262 | 84 | |
BC028725.2 | - | 2 - 3 | 163 | 84 |