Assay Design



Gene Transcripts

Gene Symbol
TCTN2
Entrez Gene ID
79867
Gene Name
tectonic family member 2
Gene Aliases
C12orf38, JBTS24, MKS8, TECT2
Chromosome Location
Chr.12: 123671108 - 123708405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024809.4 NP_079085.2 3 - 4 398 61
XM_005253623.2 XP_005253680.1 3 - 4 398 61
XM_006719605.3 XP_006719668.1 3 - 4 398 61
GenBank mRNA AK056924.1 - 3 - 4 396 61
BC009112.1 - 3 - 4 367 61
DQ278870.1 - 3 - 4 396 61

View Full Product Details