Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178034.3 | NP_828848.3 | 15 - 16 | 1674 | 58 |
XM_011521467.2 | XP_011519769.1 | 14 - 15 | 1505 | 58 | |
GenBank mRNA | AB090876.1 | - | 15 - 16 | 1677 | 58 |
BC034571.1 | - | 1 - 2 | 111 | 58 |