Assay Design



Gene Transcripts

Gene Symbol
PLA2G4D
Entrez Gene ID
283748
Gene Name
phospholipase A2 group IVD
Gene Aliases
cPLA2delta
Chromosome Location
Chr.15: 42067562 - 42094559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178034.3 NP_828848.3 4 - 5 476 69
XM_011521467.2 XP_011519769.1 3 - 4 307 69
GenBank mRNA AB090876.1 - 4 - 5 479 69

View Full Product Details