Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024109.3 | NP_077014.3 | 4 - 5 | 866 | 98 |
XM_005255570.3 | XP_005255627.1 | 4 - 5 | 866 | 98 | |
XM_005255571.2 | XP_005255628.1 | 4 - 5 | 866 | 98 | |
XM_011522657.2 | XP_011520959.1 | 4 - 5 | 866 | 98 | |
XM_017023659.1 | XP_016879148.1 | 4 - 5 | 866 | 98 | |
XM_017023660.1 | XP_016879149.1 | 5 - 6 | 1062 | 98 | |
XM_017023661.1 | XP_016879150.1 | 4 - 5 | 866 | 98 |