Assay Design



Gene Transcripts

Gene Symbol
FOXO6
Entrez Gene ID
100132074
Gene Name
forkhead box O6
Gene Aliases
-
Chromosome Location
Chr.1: 41361931 - 41383591 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291281.2 NP_001278210.2 - 49 99

View Full Product Details