Assay Design



Gene Transcripts

Gene Symbol
THAP11
Entrez Gene ID
57215
Gene Name
THAP domain containing 11
Gene Aliases
CTG-B43a, CTG-B45d, HRIHFB2206, RONIN
Chromosome Location
Chr.16: 67842310 - 67844195 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020457.2 NP_065190.2 1 - 1 1786 103
GenBank mRNA BC012182.1 - 1 - 1 1745 103
L10378.1 - 1 - 1 1620 103
L10379.1 - 1 - 1 1620 103

View Full Product Details