Assay Design



Gene Transcripts

Gene Symbol
ANKS1B
Entrez Gene ID
56899
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
Chromosome Location
Chr.12: 98729904 - 99984727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204065.1 NP_001190994.1 4 - 5 632 65
NM_001204066.1 NP_001190995.1 5 - 6 499 65
NM_001204069.1 NP_001190998.1 5 - 6 948 65
NM_001204079.1 NP_001191008.1 4 - 5 635 65
NM_181670.3 NP_858056.2 6 - 7 1128 65
XM_006719504.3 XP_006719567.1 20 - 21 3633 65
XM_006719505.3 XP_006719568.1 20 - 21 3636 65
XM_006719506.3 XP_006719569.1 20 - 21 3633 65
XM_006719507.3 XP_006719570.1 20 - 21 3636 65
XM_006719508.3 XP_006719571.1 20 - 21 3636 65
XM_011538571.2 XP_011536873.1 20 - 21 3534 65
XM_017019651.1 XP_016875140.1 20 - 21 3636 65
XM_017019652.1 XP_016875141.1 20 - 21 3624 65
XM_017019653.1 XP_016875142.1 20 - 21 3633 65
XM_017019654.1 XP_016875143.1 20 - 21 3621 65
XM_017019655.1 XP_016875144.1 20 - 21 3633 65
XM_017019656.1 XP_016875145.1 20 - 21 3531 65
XM_017019659.1 XP_016875148.1 6 - 7 1127 65
XM_017019661.1 XP_016875150.1 7 - 8 838 65
XM_017019662.1 XP_016875151.1 7 - 8 971 65
XM_017019663.1 XP_016875152.1 6 - 7 1022 65
GenBank mRNA AK289768.1 - 6 - 7 1093 65
AK295988.1 - 4 - 5 605 65
AK315954.1 - 4 - 5 487 65
AM980945.1 - 4 - 5 491 65
BC026313.1 - 6 - 7 1023 65
BC068451.1 - 5 - 6 499 65
BC150204.1 - 5 - 6 895 65

View Full Product Details